Would you like to log in again?
Sapphire Single and Multichannel Pipettes
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sapphire
89000010 89012300 89000020 89008300 89008200 89000100 89000820 89000810 89012200 89000002 89001220 89000200 89001210 89000500 89001000 89010000
76299-828CS 381.51 USD InStock
76299-828 76299-838 76299-848 76299-836 76299-824 76299-846 76299-834 76299-856 76299-822 76299-844 76299-832 76299-842 76299-852 76299-830 76299-840 76299-850
Sapphire Single and Multichannel Pipettes
Pipettes Micropipettes
Sapphire pipettes are a variable volume range of air-displacement pipettes designed and built to deliver optimal performance, ergonomics, and robustness in day-to-day use.
  • Unique serial number
  • Fully autoclaveable
  • Available in single and multichannel models with a volume range from 0.2 µl to 10 ml
  • Ergonomic design for optimum comfort
The lightweight design coupled with a state-of-the-art mechanism reduces pipetting forces and makes our pipettes both comfortable and easy-to-use. The contoured body shape allows for a relaxed grip whatever size your hand, and the 3-position tip ejector button makes pipetting comfortable for both left-handed and right-handed users.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty