Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® T Balances
Supplier: VWR
VWR®
VWR-363TC VWR-520TC VWR-1003TC VWR-10102TC VWR-105TC VWR-603TC VWR-4502TC VWR-205TC VWR-3003TC VWR-310TC VWR-2102TC VWR-8102TC VWR-160TC VWR-220TC VWR-6002TC
76300-072EA 3699.64 USD InStock
76300-072 76446-798 76300-082 76300-070 76446-800 76446-802 76300-076 76446-804 76300-074 76446-806 76300-078 76446-808 76300-080 76446-794 76446-796
VWR® T Balances
Balances and Scales
The VWR® T-Series balances feature state-of-the-art technology to provide the highest accuracy measurements.
  • Large, 5" color touch screen
  • Touchless operation via IR proximity sensors
  • Internal and external calibration
  • Seven databases
  • 2× RS232, USB (A and B), ethernet and Wi-Fi communication interfaces
  • Tool-free assembly and disassembly of the three-door draft shield
  • 5-year warranty
These balances provide accuracy and precision for any laboratory environment. Featuring internal calibration, IR proximity sensors for hands free operation, a multitude of communication options, and built in data protections, and provides advanced features for any application. The large, color touch screen allows for easy viewing of results and navigation through the balances menus. Data safety is provided via the internal memory allows for recording up to 500000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. With its 5-year warranty, this balance will provide reliable results for years to come.
Certifications: C-UL (US) and CE.
Ordering information: Accessories not included (must be ordered separately).
Delivery information: The 0.01 and 0.1 mg readability models feature a draft shield (shield can't be removed). The 0.001 g readability feature a removable, glass shield. All units supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Includes five-year warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty