Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

METTLER TOLEDO® Excellence Benchtop Density Meters, Models D4, D5 and D6

Supplier: Mettler-Toledo
30474901 30474902 30474900
76300-948EA 18790 USD InStock
76300-948 76300-950 76300-952
METTLER TOLEDO® Excellence Benchtop Density Meters, Models D4, D5 and D6
Density Meters
Developed for a wide range of applications, the Excellence Density Meters are the next generation of density measurement.

  • Color touchscreen with 7" display
  • Compatible with nine languages
  • Accuracy up to six digits
  • One Click user interface
  • BubbleCheck automatic viscosity correction and temperature control
  • Simultaneous determination of density, refractive index, pH / conductivity and color

The new space-saving Excellence Density line guarantees high reliability and maximum precision. Measure density, specific gravity, as well as many related values such as % alcohol and Brix quickly and accurately.

Ordering information: Can be further enhanced with the following Optional modules and accessories to fit your process needs: powerful autosamplers, LabX PC software, ErgoSens infrared motion sensor, LevelSens waste sensor, LogStraight fingerprint reader, SPR200 sampling pump. Contact your Avantor representative for more information.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty