Would you like to log in again?
VWR® Media Bottles, Square, PET, Sterile
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
PET250-T PET500 PET1000-T PET500-T PET250 PET1000
76299-562CS 304.56 USD InStock
76299-562 76302-840 76299-560 76302-838 76302-836 76299-564
VWR® Media Bottles, Square, PET, Sterile
Bottles Plastic Bottles
Square PET media bottles are designed for packaging, storing, or transporting powder and liquid samples.
  • Square design allows efficient use of storage space
  • PET resin is lightweight, break-resistant, and transparent
  • Bottles feature molded-in graduations
  • EB Sterilization Sterility Assurance Level (SAL) of 10⁻⁶ (ISO 11137)
  • Non-pyrogenic
VWR® Square PET Media Bottles are manufactured in a class 100000 cleanroom environment. Manufactured under EN ISO 13485:2016.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty