Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sorvall™ BIOS 16 Large Capacity Centrifuges (BIOProcessing), 240 V

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Sorvall™
75003345 75003342 75003346 75003344 75003341 75007685 75007696 75003343
76417-266PK 871.12 USD InStock
76417-266 76417-268 76417-270 76302-938 76417-272 76302-936 76417-262 76417-264
Sorvall™ BIOS 16 Large Capacity Centrifuges (BIOProcessing), 240 V
Centrifuges Floor-Standing Centrifuges
Proven productivity. Enhanced capacity of up to 8×2000 ml bottles (16 L) and the user-friendly design of large capacity centrifuges make working with large volumes easier and more convenient using the protocols you've already validated. Enhanced ergonomics and compact design combined with the quick set-up of traceable, reproducible runs makes these the centrifuges of choice for high throughput bioprocessing productivity.

  • 240 V models
  • High-throughput sample processing
  • Built-in safety and efficiency
  • Small footprint and enhanced ergonomics with push-button convenience optimize everyday centrifuge use
  • Automatic front-to-back centrifuge door opening and closing with the Auto-Door™ function
  • Auto-Lid™ rotor lid opening and closing with lid storage
  • Pre-cooling function
  • ACE integrator
  • Operational features include at start, at speed, timed start

Achieve reproducible results with the Accumulated Centrifugal Effect (ACE™) function, automatically adjusting run time to account for variations in acceleration.

The Centri-Touch™ interface with bright, highly visible and glove-friendly display simplifies run set-up, further enhanced with access controls such as user login with password protection.

It has immediate rotor detection and programming with Auto-ID instant rotor identification to save time and protect the integrity of the samples.

Support a broad array of applications and methods with the flexibility to select higher capacity rotors or match existing workflows. Choose between 4 rotors and swinging bucket sets with the option to upgrade to longer lasting Dura-Coat™ nickel-coated buckets: 6×1000 ml swinging bucket rotor; 8×1000 ml swinging bucket rotor; 6×2000 ml swinging bucket rotor or 8×2000 ml swinging bucket rotor.

This centrifuge is designed for high quality harvesting, separation and purification bioprocessing methods that require run to run reproducibility and traceability. With an enhanced capacity of up to 16 L of sample volume, a compact footprint and innovative features to optimize operations, maximizing productivity and efficiency is simplified.

Certifications: UL, CE. EN 61010-1 3rd Edition, IEC 61010-2-020 2nd Edition and IEC 61010-2-020 3rd Edition, EN 61326-1 Class B.

Ordering information: Accessories not included. Accessories/rotors must be purchased separately. Contact your Avantor representative for more information or a quote.

Delivery information: Unit will be installed by a local Thermo Scientific Represetative and training by your local specialist within five weeks.

Caution: 240 V models.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty