Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sorvall™ BIOS A Large Capacity Centrifuges (BIOProcessing), 200 to 240 V

Supplier: Thermo Scientific
Sorvall™
75007700 75007698
76302-940EA 59433.76 USD InStock
76302-940 76302-942
Sorvall™ BIOS A Large Capacity Centrifuges (BIOProcessing), 200 to 240 V
Centrifuges Floor-Standing Centrifuges
Enhanced capacity of up to 10×1000 ml (10 l in a fixed angle rotor) and the user-friendly design of large capacity centrifuges make working with large volumes easier and more convenient using the protocols you've already validated. Enhanced ergonomics and compact design combined with the quick set-up of traceable, reproducible runs makes these the centrifuges of choice for high throughput bioprocessing productivity.

  • 200 to 240 V
  • High performance for application flexibility
  • Built-in safety and efficiency
  • Small footprint and enhanced ergonomics with push-button convenience optimize everyday centrifuge use
  • Automatic front-to-back centrifuge door opening and closing with the Auto-Door™ function
  • Auto-Lid™ rotor lid opening and closing with lid storage
  • Latest global safety and compliance standards
  • Certified by UL and CE
  • Pre-cooling function
  • ACE integrator
  • Operational features include at start, at speed, timed start

Process up 10×1000 ml and up to 12000×g with the Sorvall BIOS A centrifuge and the Fiberlite™ F6-10×1000 LEX rotor, providing the benefits of carbon fiber design including corrosion and fatigue resistance and backed by a 15-year warranty.

It achieves reproducible results with the Accumulated Centrifugal Effect (ACE™) function, automatically adjusting run time to account for variations in acceleration. Centri-Touch™ interface with bright, highly visible and glove-friendly display simplifies run set-up, further enhanced with access controls such as user login with password protection.

The immediate rotor detection and programming with Auto-ID instant rotor identification to save time and protect the integrity of your samples.

Sorvall BIOS A centrifuge is designed for high quality harvesting, separation and purification bioprocessing methods that require run to run reproducibility and traceability. With an enhanced capacity of up to 10 Liters of sample volume, a compact footprint and innovative features to optimize operations, maximizing productivity and efficiency is simplified.

Certifications: UL, CE. EN 61010-1 3rd Edition, IEC 61010-2-020 2nd Edition and IEC 61010-2-020 3rd Edition, EN 61326-1 Class B.

Ordering information: Accessories not included. Must purchase accessories/rotors separately. Contact your Avantor representative for more information or a quote.

Delivery information: Unit will be installed by a local Thermo Scientific Represetative and training by your local Specialist within five weeks.

Caution: 200 to 240 V model.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty