Would you like to log in again?
Argos Technologies® PolarSafe® EVA Foam Ice Buckets and Pans, Cole-Parmer
Supplier: Cole Parmer
PolarSafe™
04394-06 04393-78 04393-60 04393-57 04394-09 04393-76 04393-93 04393-54 04394-02 04393-64 04393-88 04393-81 04393-69 04393-66 04393-90
76303-002EA 148.84 USD InStock
76303-002 76303-004 76303-010 76303-012 76303-000 76302-988 76302-998 76302-986 76302-996 76302-984 76302-994 76302-992 76302-990 76303-006 76303-008
Argos Technologies® PolarSafe® EVA Foam Ice Buckets and Pans, Cole-Parmer
Ice Containers
Mess-free superior insulation for any cryogenic medium.
  • Ethylene vinyl acetate is leak proof and will not sweat
  • Thick, nontoxic foam is compatible with a variety of cryogenic media
  • Ergonomic handles on bases and lids are glove friendly
  • Pour spouts make cleanup easy
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty