Would you like to log in again?
SP Bel-Art H-B DURAC® Calibrated Electronic Thermometers with Stainless Steel Probe, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
B60900-3000 B60900-3400 B60900-3500
76303-076CS 794.37 USD InStock
76303-076 76303-074 10753-406
SP Bel-Art H-B DURAC® Calibrated Electronic Thermometers with Stainless Steel Probe, Bel-Art Products, a part of SP
Thermometers Digital Thermometers
Monitor the temperature of pharmaceuticals, vaccines, chemicals, blood and blood products, food products and more with these high accuracy digital thermometers with chemical resistant probes.
  • Features available include 10-event recording, minimum/maximum temperature readings, timers and alarms
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish)
  • Individually serialized, CE marked
Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty