Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Traceable® 100-Hour Timer
Supplier: VWR
VWR®
90012-77
76304-420EA 39.7 USD InStock
76304-420
VWR® Traceable® 100-Hour Timer
Timers Digital Timers
100-hour timer has four countdown channels, time-of-day clock, and stopwatch.
  • Timing capacity to 99 hours, 59 minutes and 59 seconds
  • TIME IS UP flashes on display during count-up mode
  • Alarm sounds for 60 seconds and may be silenced manually
Unique 'thru-zero' allows counting down, alarming, and counting up. Time-out mode stops and starts operation. Extra-large LCD is readable from 1.8 m (6'). Quartz crystal is accurate to 0.01%. Alarm can be turned off manually or shut off automatically after one minute. 'TIME IS UP' message flashes when time reaches zero. Housed in rugged, chemical-resistant case. Unit incorporates a magnetic back, flip-open easel, spring fastener for clipping to coat, and a lanyard hole.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a two-year, 1.5 V silver-oxide battery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty