Would you like to log in again?
SP Bel-Art H-B Bluetooth® Verification, Probe and Stem Thermometers, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
B61011-5000 B60910-5000 B60910-0000
76305-142CS 901.07 USD InStock
76305-142 76305-144 76305-146
SP Bel-Art H-B Bluetooth® Verification, Probe and Stem Thermometers, Bel-Art Products, a part of SP
Thermometers Digital Thermometers
Remotely monitor temperature by broadcasting readings to any Apple or Android smartphone or device with Bluetooth® 4.0 or better utilizing the free THERMSmart app.
  • Monitor critical environments without a physical connection
  • Maximum signal distance is approximately 40 meters
  • Alarm sounds to alert user if temperature falls outside of the set parameters
  • Alarm and timer settings savable for reuse
  • LCD screen for on-unit monitoring
  • App can trigger indicator light for unit identification
  • Indicators include active alarm state and low battery
  • Individually serialized
Verification model provides accurate temperature reading without sample contamination by using a glass bead filled bottle (included) which mimics the sample.
Certifications: CE marked and RoHS compliant. Includes Certificate of Compliance indicating accuracy for 4 temperature points.
Delivery information: Batteries, multi-language (English, French, German, Italian, Portuguese and Spanish) instructions and countdown timer function are included.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty