Would you like to log in again?
HDAC-5 Inhibitor Screening Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB283359-100TEST
76305-290EA 1149.11 USD InStock
76305-290
HDAC-5 Inhibitor Screening Kit (Fluorometric), BioVision, Inc.
Assays Protein Assays
HDAC-5 Inhibitor screening kit is useful for the identification of potential HDAC-5 inhibitors.
  • Simple and reliable test to screen/study/characterize HDAC5 inhibitors
  • High-throughput compatible
  • Includes Inhibitor Control (Trichostatin A)
Histone deacetylase 5 or HDAC-5 belongs to Class II histone deacetylase family. It represses transcription by deacetylating lysine residues on the N-terminal part of histones H2A, H2B, H3 and H4. This epigenetic repression caused by HDAC-5, plays a role in transcriptional regulation, cell cycle progression and development. It interacts with myocyte enhancing factor (MEF-2) proteins that are associated with colon cancer, causing the repression of MEF-2 dependent gene expression.
HDAC-5 is also involved in epigenetic regulation of ESR1 expression in breast cancer. HDAC-5 inhibition induces reactive oxygen species (ROS) production, ultimately leading to apoptotic cell death as well as mechanisms of mitochondrial quality control processes such as mitophagy and mitobiogenesis. Therefore, HDAC-5 inhibition in cancer cells significantly reduces tumor size due to apoptotic cell death of cancer cells (Hendrick, et al, Oncogene. 2017 Aug 24; 36(34):4859-4874), which makes HDAC-5 an attractive target for anti-cancer therapy.
BioVision’s HDAC-5 Inhibitor screening kit is a plate-based fluorometric kit for screening potential HDAC-5 inhibitors. The principle of the kit is based on the deacetylation of an acetylated substrate by HDAC-5. The deacetylated product is then cleaved by a developer to release a highly fluorescent moiety with Ex/Em= 380/460 nm. Trichostatin A, a well-known HDAC inhibitor is provided as inhibitor control.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty