Would you like to log in again?
Rectangular and Square Cover Slips, Bellco Glass
Supplier: Bellco Glass
1916-09035A 1916-09022 1916-10522A 1916-13054A 1916-10550A 1916-09009A 1916-25075 1916-24050A 1916-10535A 1916-09050A 1916-12222A
76305-492CS 646.95 USD InStock
76305-492 76305-508 76305-494 76305-496 76305-498 76305-500 76305-488 76305-502 76305-504 76305-490 76305-506
Rectangular and Square Cover Slips, Bellco Glass
Cover Glasses
These cover slips provides good quality, reliability, and better performance for all laboratory microscopy needs
  • #1 thickness (0.13 to 0.16 mm)
  • Packed 100 per case
Clear white borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty