Would you like to log in again?
FLASK NEPHLO BOROSILICAGE GL 300ML CS4
Supplier: Bellco Glass
Flask Nephlo Borosilicage Gl 300Ml CS4^ Nephlo Flask, Manufactured From Type 1 Class A Borosilicage Glass With Cleanout Port And Depressed Sidearm. Supplied With Black Phenolic Rubber Lined 38-415 Center Cap And 18-415 Sidearm Cap. Sidearm Dimensions: 12
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FLASK NEPHLO BOROSILICAGE GL 300ML CS4 76305-588 2574-12133 Case of 4 (300ml) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty