Would you like to log in again?
Monarch® RNA Cleanup Columns, New England Biolabs
Supplier: New England Biolabs
Monarch®
T2037L T2037L
76307-362EA 264 USD InStock
CA76307-362 76307-362
Monarch® RNA Cleanup Columns, New England Biolabs
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Monarch® RNA cleanup columns (10 µg) are a component of the Monarch RNA cleanup kit (10 µg) and can be used to purify up to 10 µg of RNA from enzymatic reactions. They are supplied with Monarch® collection tubes II. RNA cleanup binding buffer and Monarch® RNA wash buffer, are also available separately.
  • Optimal for cleaning up RNA from a variety of enzymatic reactions
  • Ideal for the concentration of RNA into small volumes (elute in ≥6 µl)
  • 70 to 100% RNA recovery, even with inputs <20 ng
  • Purified RNA is ready for use in a wide variety of downstream applications
  • Unique column design prevents buffer carryover and elution of silica particulates
The columns are designed without the use of a retaining ring, ensuring no buffer retention and no carryover of contaminants. RNA can be eluted in as little as 6 μl and is ready for use in a variety of downstream applications including RT-PCR, RNA library prep for NGS, and labeling.
Monarch® RNA cleanup columns are also available for 50 and 500 µg binding capacities.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty