Would you like to log in again?
BALANCE ML104T/A ANALYTICAL TRADE LEGAL
Supplier: Mettler Toledo
Balance Ml104T/A Analytical Trade Legal^ Analytical Balance Ml104T/A00, Repeatability (Test Wt): 0.1 Mg (100 G), Minimum Wt (Usp, 0.1%, Typical): 0.16 G, Display: 4.5In Color Tft Touchscreen, Dimensions: 290Mmx331Mmx 193Mm, Applications: Backweighing; Counting; Check Weighing, Maximum Capacity: 120G
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BALANCE ML104T/A ANALYTICAL TRADE LEGAL 76307-372 30243395 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty