Would you like to log in again?
BALANCE MS403TS/A PRECISION TRADE LEGAL
Supplier: Mettler Toledo
Balance Ms403Ts/A Precision Trade Legal^ Precision Balance Ms403Ts/A00, Repeatability : 0.7 Mg, Minimum Wt (Usp, 0.1%, Typical): 1.4 G, Display: 7 In Color Tft Touchscreen, Dimensions: 347 Mm X 283 Mm X 204 Mm, Applications: Back-Weighing; Check Weighing; Counting; Density; Maximum Capacity: 420 G
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BALANCE MS403TS/A PRECISION TRADE LEGAL 76307-408 30133542 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty