Would you like to log in again?
BALANCE MS6002TS/A PRECISION TRADE LEGAL
Supplier: Mettler Toledo
Balance Ms6002Ts/A Precision Trade Legal^ Precision Balance Ms6002Ts/A00, Repeatability(Typical) : 0.01 G (2,000 G), Minimum Wt (Usp, 0.1%, Typical): 14 G, Display: 7 In Color Tft Touchscreen, Dimensions: 347Mm X 99Mm X 194Mm, Applications: Backweighing; Counting; Density; Maximum Capacity: 6200 G
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BALANCE MS6002TS/A PRECISION TRADE LEGAL 76307-422 30133549 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty