Would you like to log in again?
BALANCE MS16001L/A03 PRECSION TRADELEGAL
Supplier: Mettler Toledo
Balance Ms16001L/A03 Precsion Tradelegal^ Balance Precsion Tradelegal Ms16001L/A03, Weighing Pan Diameter: 245 X 351 Mm, Fully Automatic Time- And Temperature-Controlled Internal Adjustment (Fact) And Robust Monobloc High-Performance Weighing Cell, Assured Of Strong Performance, Size: 16200 X 1 G
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BALANCE MS16001L/A03 PRECSION TRADELEGAL 76307-432 11145017 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty