Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Rebel Hybrid Microscope, Upright and Inverted, Discover Echo
Supplier: ECHO
Rebel
RCND3013 RHDR0002 RCND3002 RCND3001 RHDR0006 RSTG3001 RSTG1001 RMAG0113 RBLT3000
76376-762EA 265.43 USD InStock
76376-762 76376-750 76376-752 76376-754 76307-596 76376-746 76376-756 76307-594 76307-592
Rebel Hybrid Microscope, Upright and Inverted, Discover Echo
Microscopes
Rebel is designed around the user to challenge the status quo. Designed from the ground up to improve microscopy.
  • ECHO Pro: powerful yet simple software reimagined to collaboratively view, capture, and share images
  • Ultra high resolution 13" touch display
  • CMOS color camera (8 MP)
  • Plan Achromat and Apochromat options available
  • XY mechanical stage option available
  • Ergonomic biosafety cabinet and adjustable height table options available
Versatility: Hybrid microscope easily converts between upright and inverted configurations. Brightfield and phase contrast modes available. Eyepiece option available as well.
Productivity: Our award-winning touch interface is incredibly easy to use. Eyepieces have been replaced with a 13" iPad Pro with Retina Display to view and capture stunning crystal-clear images.
Rebel True Ergo Table and Rebel Logic + Biosafety cabinet by Labconco bring the microscope stage down flush to the tabletop, setting a new precendent in microscope ergonomics.
Ordering information: Microscope includes a one-year warranty on electrical, and five-year warranty on mechanical parts and optics. Contact your VWR representative to demo the the newest innovations in microscopy. A Rebel Hybrid Microscope Bundle option is also available (see 76359-898).
Delivery information: Includes microscope body, iPad Pro with locking stand, 8mp CMOS color camera, power cord, microscope cover, and lens cloth.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty