Would you like to log in again?
SureSTART™ Glass Screw Top Vials, 2 ml, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SureSTART™
CHSV9-10P CHSV9-10PF CHSV9-10PT CHSV9-20PF CHSV9-20P
76307-600PK 19.62 USD InStock
76307-600 76307-598 TSCHSV9-10PF TSCHSV9-10PT TSCHSV9-20PF
SureSTART™ Glass Screw Top Vials, 2 ml, Level 1 Everyday Analysis, Thermo Scientific
Vials
Use performance level 1 Thermo Scientific™ SureSTART™ 2 ml glass screw top vials for everyday chromatography analysis. They provide an affordable choice and can be used with all GC and HPLC/UHPLC instrument types. These 2 ml glass screw top vials are specification certified to ensure autosampler compatible physical dimensions. Choose from clear and amber glass. The vials meet all requirements of US, EU, and JPN Pharmacopia. Seal these screw top vials with 9 mm screw caps and septa.
  • Performance level 1 vials – for everyday analyses
  • Have a single wave on the vial
  • Are suitable for QA/QC applications using core detectors
  • For use to verify compound presence and quantify using detection techniques such as UV, DAD/PDA, RI, Fluorescence, ELSD, FID, NPD, and ECD
  • Suitable for GC and HPLC/UHPLC applications
  • Convenient write-on patch on the vials
About Thermo Scientific SureSTART vials, caps, and kits:
Thermo Scientific SureSTART vials and caps are available in three performance levels. Now you can choose the products that best suit your analytical, performance, and budget needs.
- Level 1 vials and caps are for everyday applications.
- Level 2 vials, caps, and kits are for high throughput applications. These vials have very low metal content, extractables, and leachables to deliver high quality data with an uninterrupted workflow.
- Level 3 vials, caps, and kits are for high performance applications. Use level 3 for your most valuable samples (and analytes prone to glass adsorption) to deliver consistent, reproducible results for your most demanding analyses.
Certifications: Specification certified. All Thermo Scientific SureSTART™ vials are dimensionally verified, Inspected by automated optical and gauge inspection to ensure critical dimensions and tolerances, Uniform and defect-free, Lot traceable, 100% traceable through the manufacturing process, Certificate of Conformance available by lot number upon request.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty