Would you like to log in again?
VAI® WipeDown Dry Wipe, Veltek Associates
Supplier: Veltek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VAI®
VEL8-12X12-S-3002 VEL8-12X12-S-3031
76308-434CS 1270.93 USD InStock
76308-434 89204-832
VAI® WipeDown Dry Wipe, Veltek Associates
Wipes Cleanroom Wipes Dry Cleanroom Wipes
WipeDown is designed as a dry cleaning wipe for use during wipe downs of filling and packaging machinery, stainless steel surfaces, lexan, polycarbonate, glass and any other critical surface that requires cleaning.
  • Laundered polyester
  • Laser cut sealed edge
  • Low particulate and low extractable
These dry wipes are constructed of a 100% continuous polyester fiber that is designed to be clean and absorbent.
VAI®'s dry wipes are ultra-clean, superior, laundered dry wipes suitiable for use in a cleanroom environment. Specifically designed to be clean and absorbent, using FocusEdge cutting technology to cut wipes that seal the edges and reduce shedding to a minimum during wipe down or general use.
Avaliable in two sorbancy options. WipeDown Dry Wipes are ideal for use in both aspetic and non-aseptic wipe downs on any critical surface that requires cleaning. WipeDown HighSorb Dry Wipes are ideal for use in both aseptic and non-aseptic areas to keep the surface dry, residue-free, and streak-free after chemical application.
Packaging: Each sterile bag is bulk packaged in an additional single outer bag pack.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty