Would you like to log in again?
Brain Heart Infusion Broth, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
C5143 C5141 C5140 C5140 C5142 C5144 C5142 C5143 C5141
89405-138EA 182.4 USD InStock
CA89405-140 CA89405-142 89405-138 76308-444 89405-136 CA89405-136 CA89405-138 89405-142 89405-140
Brain Heart Infusion Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Brain Heart Infusion Broth is a general purpose nutrient medium recommended for the cultivation and isolation of a variety of fastidious and nonfastidious microorganisms, including bacteria, yeasts, and mold
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty