Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CellXpert® C170 CO₂ Incubators

Supplier: Eppendorf
CellXpert®
2231000759 6734010045 6734010045 2231000759 6734010015 6734010015
77321-656EA 11919.36 USD InStock
77321-656 CA77321-656 76308-822 MPCA76308-823 76460-802 CA76308-822
CellXpert® C170 CO₂ Incubators
Incubators CO₂ Incubators
This CO₂ incubator features a highly uniform atmosphere, fast recovery and effective contamination protection. A device that is easy to clean and use with a high usable volume to footprint ratio.

  • Easy cleaning: Seamless chamber, easy disassembly in 40 seconds
  • Effective contamination protection: 140 °C High temperature disinfection (HTD), no internal fan-associated HEPA-filters necessary
  • Temperature uniformity: Innovative heating concept with multiple independent temperature sensors - verified at 27 spots inside the incubator according to German DIN 12880 norm
  • Fast recovery after door opening: Temperature and CO₂ in less than five minutes - no regulation overshoot for sample safety
  • Enhanced vibration and turbulence protection: No fan inside
  • Save money: Up to 25% more usable space by fanless design, no internal expendable parts (e.g. fan-associated HEPA-filters), significantly reduced gas consumption

Easy to handle the new magnetic door locking concept for outer and inner doors. It enhanced vibration and turbulence protectes the fan-less design. Easy cleaning and disinfection, seamless chamber and easy disassembling.

Features uniform temperature verified at 27 points (German Norm DIN 12880) by multiple temperature sensors and four individually controlled heating circuits. No internal HEPA-filters that potentially contaminate the chamber after HTD.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty