Would you like to log in again?
HyFlex® 11-928 Medium-Duty Industrial Gloves, Ansell
Supplier: Ansell
HyFlex®
11928090 11928110 11928080 11928100 11928070
76308-892CS 1901.3 USD InStock
76308-892 76308-894 76308-926 76308-922 76308-924
HyFlex® 11-928 Medium-Duty Industrial Gloves, Ansell
Gloves
INTERCEPT™ technology ANSI cut level 4 performance for the ultimate in cut resistance.
  • Glass-free INTERCEPT™ liner provides increased cut protection
  • Silicone and DMF-free
  • Grip performance for safe handling in dry to light oily applications
  • Double dipped nitrile coating protects against oil and industrial fluid exposure
  • INTERCEPT™ Aramid Liner for increased cut performance
  • Contact Heat Protection level 1
The glove is composed of double layer RIPEL™ nitrile coating, which keeps hands dry and comfortable when handling oily parts and tools. The HyFlex® 11-928 also offers intermittent contact hear protection when handling hot parts up to 212 °F.
Certifications: ANSI 3 ABR; ANSI A4 CUT; EN 420:2003 + A1:2009; Category lll; REACH compliant.
Packaging: Gloves are packaged 12 pairs per polybag, 12 polybags per carton.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty