Would you like to log in again?
HiScale Columns Packed with Superdex Increase or Superose Increase Resins, Cytiva
Supplier: Cytiva
HiScale™
29321903 29-3219-08 29321905 29-3219-04 29-3219-03 29-3219-06 29321904 29-3219-07 29321908 29321907 29-3219-05 29321906
CA76308-950EA 17690.33 USD InStock
CA76308-950 76308-974 76308-952 CA76308-974 76308-976 CA76308-952 76308-950 76308-948 CA76308-948 76308-978 CA76308-976 CA76308-978
HiScale Columns Packed with Superdex Increase or Superose Increase Resins, Cytiva
Chromatography Columns
These HiScale columns packed with Superdex Increase or Superose Increase resins are used to purify larger volumes of up to 8.5 ml.
  • High-resolution SEC columns: Small particle size and narrow particle size distribution of the packed resin ensures highly pure protein
  • HiScale columns packed with SEC resins on demand, please request a quote
HiScale columns are packed on demand for you with Superdex 200 Increase, Superdex 75 Increase, and Superose 6 Increase SEC (gel filtration) resins. The columns are available in two sizes: 16 × 400 mm and 26 × 400 mm. These two column sizes enable purification of sample volumes up to 3.2 and 8.5 ml, respectively.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty