Would you like to log in again?
CT™ Flange Plug Caps for CapTrack™ Systems, Globe Scientific
Supplier: Globe Scientific
CT™
CT-CAP-B CT-CAP-K CT-CAP-N CT-CAP-Y CT-CAP-G CT-CAP-R CT-CAP-W CT1-CAP CT-CAP-L CT-CAP-C
76309-002PK 31.4 USD InStock
76309-002 76308-996 76309-000 76308-998 76309-006 76308-992 76309-004 76308-994 76309-008 76308-990
CT™ Flange Plug Caps for CapTrack™ Systems, Globe Scientific
Closures Test/Sample Tube Closures
CT™ flange plug caps have extremely high tolerances and are specifically designed for use with CapTrack™ systems.
  • Low density polyethylene (LDPE)
  • Assorted colors available
  • Fits both glass and plastic tubes
These caps provide superior leak resistance in all tubes processed by the CapTrack™ benchtop recapping archiver, which allows for the utmost confidence in sample integrity during handling and storage of specimens.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty