Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD274 Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-101-GU8S 600-101-GU8
76309-332EA 518 USD InStock
76309-332 76309-334
Anti-CD274 Goat Polyclonal Antibody
Antibodies
PDL1 belongs to the immunoglobulin superfamily and BTN/MOG family. PDL1 is involved in the costimulatory signal, essential for T-cell proliferation and production of IL-10 and IFNG, in an IL-2 dependent and a PDCD1 independent manner. It is up-regulated in T-and B-cells, dendritic cells, keratinocytes and monocytes after LPS and IFNG activation. PDL1 is up-regulated in B-cells activated by surface Ig cross-linking. Interaction with PDCD1 inhibits T-cell proliferation and cytokine production. During infection or inflammation of normal tissue, this interaction is important for preventing autoimmunity by maintaining homeostasis of the immune response. In tumor microenvironments, this interaction provides an immune escape for tumor cells through cytotoxic T-cell inactivation. Expression of this gene in tumor cells is considered to be prognostic in many types of human malignancies, including colon cancer and renal cell carcinoma. Anti-PDL-1 Antibody is useful for researches interested in cancer research, NF-kB pathway research, and the Immune system.

Type: Primary
Antigen: CD274
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty