Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPK14 Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-101-HB4S 600-101-HB4
76309-336EA 518 USD InStock
76309-336 76309-338
Anti-MAPK14 Goat Polyclonal Antibody
Antibodies
p38, also called SAPK2A, belongs to the MAP kinase family. The MAP kinases are critical signaling proteins for many biochemical signals. They participate in transcription regulation, cellular differentiation and proliferation. Environmental signals and cytokines catalyze the kinases into bonding their substrates such as MAX and ATF2, for example, and p53 a tumor suppressing gene. p38 may be associated with diseases such as chlamydia and patellar tendinitis. Anti-p38 antibody is useful for researchers interested in signal pathways, cellular development or cancer research.

Type: Primary
Antigen: MAPK14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty