Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DsRed Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-379-RTU
76309-344EA 315 USD InStock
76309-344
Anti-DsRed Rabbit Polyclonal Antibody
Antibodies
Fluorescent proteins such as green fluorescent protein (GFP) or Discosoma Red Fluorescent Protein (or DsRFP) are widely used in research practice. DsRed is isolated from sea anemone Discosoma sp. mushroom. GFP and DsRed share only 19% identity, therefore, in general, anti-GFP antibodies do not recognize DsRed protein and vice versa. Structurally, DsRed protein is similar to GFP protein in terms of its overall fold (a β-can) and chromophore-formation chemistry. However, GFP protein undergoes an additional step in the chromophore maturation and obligates tetrameric structure. Using site-directed mutagenesis, several DsRed protein variants have been created allowing the red fluorescent protein mature as a monomeric form. Among DsRFP monomeric variants are monomeric mutant mRFP1, mBanana, mCherry, mHoneydew, mPlum, mOrange, mStrawberry and mTangerine that offer a wide range of fluorescent colors. As Rockland RFP polyclonal antibodies are raised against whole RFP protein of wild type, the polyclonal antibodies are expected to recognize all RFP variant forms. This RFP antibody has been pre-absorbed to eliminate any potential cross-reactivity to human, mouse and rat serum proteins. The antibodies are also confirmed for non-reactivity to GFP protein. All Rockland Immunochemical’s RFP antibodies are affinity-purified to assure both high affinity and specificity. Rigorous quality control testing ensures that the finished product meets or exceeds out high standards for optimum performance in your assays.

Type: Primary
Antigen: DsRed
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty