Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYP1A1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-HB7S 600-401-HB7
76309-354EA 148 USD InStock
76309-354 76309-352
Anti-CYP1A1 Rabbit Polyclonal Antibody
Antibodies
Cytochrome p450 belongs to an enzyme superfamily that catalyzes many reactions such as the synthesis of cholesterol, lipids, steroids by oxidizing the compounds and plays a role in drug metabolism. The protein has been known to be stimulated by cigarette smoke. On-going studies link the cytochrome p450 protein with lung cancer. In the liver microsome, it is part of the NADPH-dependent electron transport pathway. The protein has been found expressed in the lung, lymphocytes and placenta. Anti-Cytochrome p450 antibody is useful for researchers interested in cancer research and enzyme research.

Type: Primary
Antigen: CYP1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: human, rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty