Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TLR4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-MK3 600-401-MK3S
76309-358EA 148 USD InStock
76309-358 76309-356
Anti-TLR4 Rabbit Polyclonal Antibody
Antibodies
TLR4 (Toll-Like Receptor 4). Toll-like receptors (TLRs) are signaling molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses.  These proteins act through adaptor molecules such as MyD88 and TIRAP to activate various kinases and transcription factors such as Protein Kinase C (PKC) alpha/beta and NF-kB.  Studies with TLR4-deficient mice indicate that the main ligand for TLR is lipopolysaccharide.  Consequently, these mice also showed increased susceptibility to Gram-negative sepsis. Mutations in this gene have been associated with differences in LPS responsiveness, macular degeneration, and pertussis. Anti-TLR4 Antibody is useful for researchers interested in transcription factor research, cytokine research, MIF action through Endocytic Pathway, and rheumatoid arthritis.

Type: Primary
Antigen: TLR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty