Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGB3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GU0S 600-401-GU0
76310-206EA 518 USD InStock
76310-206 76310-208
Anti-ITGB3 Rabbit Polyclonal Antibody
Antibodies
Integrin beta 3, also called ITB3, belongs to the integrin beta chain family. Integrin alpha-V/beta-3 (ITGAV/ITB3) is a receptor for cytotactin, fibronectin, laminin, matrix metalloproteinase-2, osteopontin, osteomodulin, prothrombin, thrombospondin, vitronectin and von Willebrand factor. Integrin alpha-IIb/beta-3 (ITGA2B/ITGB3) is a receptor for fibronectin, fibrinogen, plasminogen, prothrombin, thrombospondin and vitronectin. Integrins alpha-IIb/beta-3 and alpha-V/beta-3 recognize the sequence R-G-D in a wide array of ligands. ITGA2B/ITGB3 recognizes the sequence H-H-L-G-G-G-A-K-Q-A-G-D-V in fibrinogen gamma chain. Following activation integrin alpha-IIb/beta-3 brings about platelet/platelet interaction through binding of soluble fibrinogen. This step leads to rapid platelet aggregation which physically plugs ruptured endothelial surface. ITGAV/ITB3 binds to CX3CL1, NRG1, FGF1, IGF1, PLA2G2A, and FBN1. ITGAV/ITGB3 acts as a receptor for herpes virus 8/HHV-8, coxsackievirus A9, Hantaan virus, cytomegalovirus/HHV-5, human metapneumovirus, human parechovirus 1, and west nile virus. Intergrin beta 3 may be associated with disorders such as Glanzmann thrombasthenia, myocardial infarction, purpura, and insulin-like growth factor i deficency. Anti-Integrin beta 3 Antibody is useful for researched interested in RET Signaling, MAPK-Erk Pathway, ERK Signaling, and Integrin Pathway Research.

Type: Primary
Antigen: ITGB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty