Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ATF6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GU9 600-401-GU9S
76310-446EA 518 USD InStock
76310-446 76310-448
Anti-ATF6 Rabbit Polyclonal Antibody
Antibodies
ATF6 belongs to the bZIP (basic-leucine zipper domain transcription factor) family. Activating target genes for the unfolded protein response during endoplasmic reticulum stress. Embeded in the endoplasmic reticulum, it functions as an ER stress sensor/stress transducer. Following stress-induced proteolysis it functions as a nuclear transcription factor via a cis-acting ER stress response element (ERSE) that is present in the promoters of genes encoding ER chaperones. Binds DNA on the 5'-CCAC[GA]-3' half of the ER stress response element (ERSE) (5'-CCAAT-N(9)-CCAC[GA]-3') and of ERSE II (5'-ATTGG-N-CCACG-3'). Binding to ERSE requires binding of NF-Y to ERSE. ATF6 could also be involved in the activation of transcription by the serum response factor. This protein may play a role in the development of the foveal and cone function in the retina. Therefore, attributing to disorders such as achromatopsia, blindness, nystagmus, myopia, cataracts and more. Anti-ATF6 Antibody is useful for researchers studying eye development, protein folding and processing, Endoplasmic Reticulum Stress Pathways, Glucose and Energy Metabolism Research.

Type: Primary
Antigen: ATF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty