Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-POLR2A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-HB3S 600-401-HB3
76309-350EA 148 USD InStock
76309-350 76310-450
Anti-POLR2A Rabbit Polyclonal Antibody
Antibodies
Anti-POL II antibody will detect DNA-directed RNA polymerase II subunit RPB1, an enzyme composed of 12 subunits, which initiates the transcription of RNA from DNA, synthesizing many functional non-coding RNAs as well as messenger RNA precursors. POL II encodes a core element of the transcription machinery. Malfunctioning of the Pol II gene can cause diseases such as Cockayne Syndrome B and La Crosse Encephalitis. Anti-POL II Antibody is useful for researchers interested in DNA damage and repair studies.

Type: Primary
Antigen: POLR2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty