Would you like to log in again?
Laboratory Benchtop Autoclaves, Kinematica
Supplier: Kinematica
A1200010 A1220010 A1240010
76310-486EA 19869.89 USD InStock
76310-486 76310-488 76310-490
Laboratory Benchtop Autoclaves, Kinematica
Sterilizers Autoclaves (Steam Sterilizers)
These bench-top autoclaves not only fit on any laboratory bench, the chamber is also designed to achieve optimum loading capacity with many standard bottles (Schott) and Erlenmeyer flasks.
  • Programmable
  • Holds 1 l bottles
  • Self contained - no plumbing required
  • Touch screen control for greater flexibility and easy handling
Angular sterilizer chambers according to the BoTec principal, high-strength and efficient. The results are shorter loading times and energy savings at the same time. 10 years guarantee for the inner chamber and the steam kettle. Due to the angular chamber, an optimal use of the chamber capacity is achieved. Compared with a round chamber with equal chamber capacity, the loading capacity is 36% higher.
Chamber, double mantle and steam generator are made entirely of corrosion-free stainless steel, 1.4571/ AISI 316Ti. The case and support frame are made entirley of corrosion-free 1.4301/ AISI 304 stainless steel, which is resistant to all common disinfectants. Further, the case has a semi-gloss surface for easier cleaning. All pipes exposed to steam are made of 1.4404/ AISI 316L stainless steel and isolated particle-free. Pneumatically controlled steel valves for durability and low operation costs.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty