Would you like to log in again?
Notrax® 109 Brush Step® Floor Mattings, Justrite®
Supplier: Justrite
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
NoTrax®
109S0034BR 109S0034BU 109S0310BR 109S0035GN 109S0035RB 109S0023GN 109S0046RB 109S0023BU 109S0034GN 109S0046GN 109S0034RB 109S0310GN 109S0034CH 109S0035BR 109S0310RB 109S0036BU 109S0310BU 109S0036RB 109S0035CH 109S0046CH 109S0046BU 109S0048BR 109S0036GN 109S0046BR 109S0036BR 109S0048GN 109S0035BU 109S0023BR 109S0036CH 109S0023CH 109S0048RB 109S0048CH 109S0048BU 109S0023RB
76310-638EA 132.85 USD InStock
76310-638 76310-616 76310-618 76310-882 76310-586 76310-884 76310-600 76310-886 76310-888 76310-900 76310-580 76310-582 76310-560 76310-880 76310-584 76310-902 76310-596 76310-552 76310-630 76310-598 76310-554 76310-896 76310-632 76310-610 76310-556 76310-578 76310-898 76310-634 76310-612 76310-558 76310-636 76310-614 76310-592 76310-594
Notrax® 109 Brush Step® Floor Mattings, Justrite®
Floor Mats Permanent Floor Mats
Brush Step® Matting features a lighter-weight double rib high/low design that performs the scraping function of the mat. The ribbed surface brushes, channels, and conceals dirt and debris. It is the perfect choice for light to medium traffic areas.
  • Polypropylene carpet with vinyl backing
  • 18 ounces of needle-punched yarn per square yard
  • Low profile to perform in narrow clearance doorways
  • Vinyl backing ensures minimal buckling and mat movement
  • Ribs run the length of the mat
Ideal for indoor use in entranceways, lobbies, and all light to medium traffic areas.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty