Would you like to log in again?
Notrax® 562 Sanitop® Floor Matting, Justrite®
Supplier: SUPERIOR MANUFACTURING GROUP INC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
NoTrax®
562S0035BL
76310-654EA 97.54 USD InStock
76310-654
Notrax® 562 Sanitop® Floor Matting, Justrite®
Floor Mats Traction and Anti-Fatigue Floor Mats
Sanitop® mats are made from a durable rubber compound designed to withstand harsh chemicals while creating a slip resistant surface.
  • Durable rubber compound
  • Molded alternating large/small hole drainage system keeps moisture and debris free from work area
  • Raised pattern surrounds large holes to channel drainage and provide worker traction
  • Beveled ramps on all 4 sides
  • 15 sq. ft. coverage area
A large and small hole drainage system channels liquid and debris away from the work area. Molded-in beveled edges on all four sides eliminate trip hazards. Sanitop® offers a simple, economical solution for wet environments in many industrial or food service applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty