Would you like to log in again?
Notrax® 539 WebTrax® Floor Mattings, Justrite®
Supplier: SUPERIOR MANUFACTURING GROUP INC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
NoTrax®
539R0340GY 539S0035BU 539R0440GY 539R0340BU 539R0440BU 539S0035GY
76310-740EA 355.6 USD InStock
76310-740 76310-840 76310-742 76310-830 76310-842 76310-828
Notrax® 539 WebTrax® Floor Mattings, Justrite®
Floor Mats Traction and Anti-Fatigue Floor Mats
WebTrax® is a multi-functional vinyl floor mat that can be used in a variety of wet applications.
  • Industrial grade extruded PVC
  • Open construction of extruded vinyl has a wave design that creates a slip-resistant walking surface
  • Oil-, grease-, and UV-resistant
  • Performs well in different climates and adapts to any surface
  • Free of: DOP, DMF, silicone, heavy metal, ozone depleting substances
  • Factory compressed borders
The wave pattern design creates a slip-resistant walking surface that increases traction on wet and slippery floors. Antibacterial treated PVC material prevents the growth of fungi or bacteria while the open structure design allows for easy draining of liquids. The industrial grade PVC compound also provides greater resistance to caustic chemicals, oils, and greases.
Material PVC
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty