Would you like to log in again?
CELLVIEW SLIDE 10WELL ADVANCED TC CS20
Supplier: Greiner Bio-One
Cellview Slide 10Well Advanced Tc CS20^ Cellview Slide, 10 Well, Sterile, advanced TC, Sterile, glass bottom, 10 compartments, single packed
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CELLVIEW SLIDE 10WELL ADVANCED TC CS20 76311-042 543978 Case of 20 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty