Would you like to log in again?
Vesta-Syde® SQ 64 st. RTU Quaternary Ammonium-Based Disinfectant, STERIS®
Supplier: Steris
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1S2408 1S24Y4
76311-086CS 646.96 USD InStock
76311-086 76311-090
Vesta-Syde® SQ 64 st. RTU Quaternary Ammonium-Based Disinfectant, STERIS®
Cleaning Agents Disinfectants
Vesta-Syde SQ 64 Ready to Use Quaternary Ammonium Disinfectant is the sterile version of our phosphate-free one-step cleaner and disinfectant formulated for use on hard, non-porous surfaces. Designed to be an effective one-step disinfectant cleaner.
  • Sterile and phosphate-free
  • Ease of Use - Eliminates the need to validate dilution preparation procedures by providing a consistent dilution every time
  • Convenience - Same key ingredients as Vesta-Syde SQ Disinfectant concentrate to simplify validation requirements
  • Productivity - Low residue formulation to maintain visibly clean surfaces with minimal rinsing
  • Efficacy - Microbial control against bacteria, fungi, and viruses using AOAC and EN protocols
  • Availability - Active ingredients notified according to the Biocide Product Directive and registered with the US EPA
  • Assurance - Manufactured in accordance with the cGMP with complete lot traceability
Made with same main ingredients as the Vesta-Syde SQ Disinfectant concentrate, Vesta-Syde SQ RTU Disinfectants kill a broad spectrum of common bacteria and fungi. At a use-dilution rate of 1.6% v/v (1:64), Vesta-Syde SQ 64 Ready to Use Disinfectant requires no mixing or activation for convenient and consistent use.
The Ready to Use solution can be applied manually or sprayed.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty