Would you like to log in again?
DISINFECTANT VESTASYDE SQ128 ST 1GAL CS4
Supplier: STERIS CORP
Disinfectant Vestasyde Sq128 St 1Gal CS4^ Vesta-Syde SQ* 128 Ready-To-Use Disinfectant, Sterile, Quaternary ammonium based formula designed to be an effective 1-step disinfectant cleaner for use in pharmaceutical manufacturing facilities, Phosphate-free one-step, For use on washable hard, Size: 1gal
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DISINFECTANT VESTASYDE SQ128 ST 1GAL CS4 76311-102 1S2508 Case of 4 (1/Itm/Bx) Retrieving Restricted
Restricted To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty