Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Floortex® Cleartex® Ultimat® Polycarbonate Chair Mat for Low/Medium Pile Carpet, Essendant LLC FKA Lagasse

Supplier: Essendant

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Floortex ClearTex®
FLREC118923ER
76311-310EA 120.91 USD InStock
76311-310
Floortex® Cleartex® Ultimat® Polycarbonate Chair Mat for Low/Medium Pile Carpet, Essendant LLC FKA Lagasse
Floor Mats Permanent Floor Mats
Polycarbonate chair mat for low to medium pile carpets.

  • The ultimate in quality, clarity and durability for home and office flooring
  • Produced from 100% virgin original floortex polycarbonate for ultimate performance and durability
  • Polycarbonate does not curl, crack, discolor or smell under correct application
  • Twice the impact strength of PVC for a rigid easy glide surface for maximum ergonomic performance
  • Free of phthalates, tin, lead and cadmium content
  • Manufactured using 30% renewable energy
  • Gripper back for low/medium pile carpets up to 1/2" thick
  • 200 lbs. capacity, no lip

High clarity allows the beauty of flooring to shine through.

Certifications: GREENGUARD certified; GREENGUARD gold certifed.

Ordering information: Includes small package indicator.

Caution: This product expose to Bisphenol A (BPA), which is known to the State of California to cause birth defect/reproductive harm. For more information go to www.P65Warnings.ca.gov.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty