Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Solaris™ 2000 & 4000 Orbital Shakers, 100 to 240 V

Supplier: Thermo Scientific
Solaris™
SK1824 SK4000 SK2000 SK1818 SK1830D SK1214 SK4000 SK1214D SK1830 SK1818D SK3624
76311-670EA 454.67 USD InStock
76311-670 76311-660 76311-680 76311-678 76311-676 76311-666 76311-664 76311-672 76311-662 85022-770 76311-684
Solaris™ 2000 & 4000 Orbital Shakers, 100 to 240 V
Shakers and Mixers Orbital Shakers
The evolution of shaking. A revolution in productivity.

  • 100/240 V models
  • Exceptional reliability with heavy-duty drive mechanism
  • Flexible application support to obtain the most value
  • User programmability for easy set up and reproducibility
  • Compact footprint designed to fit on your bench-top
  • Connectivity for easy monitoring
  • Industry-leading warranty for peace of mind
  • Triple eccentric drive with maintenance-free belt system

Solaris™ Orbital Shakers set a new standard for reliability and technical innovation. Ruggedly built for demanding use, they combine a new ergonomic design with key feature enhancements that help save you time, protect your process and drive your science forward. The triple eccentric drive mechanism is built upon proven technology to easily support heavy loads. The belt is maintenance-free and engineered to last the life of the shaker, and the shaker mechanism itself comes with a 10-year warranty. Available in two convenient sizes for benchtop, Solaris™ Orbital Shakers offer a variety of platforms plus associated flask clamps and tube racks to support virtually any capacity you require. These shakers can operate in microbiological incubators, environmental chambers, or refrigerators to meet a wide range of application requirements.

Easy shaker maintenance simplifies your day. Solaris™ Orbital Shakers offer easy-to-remove platforms that make cleaning fast and easy. Featuring rounded corners, you can clean the unit quickly and completely with confidence. The sleek design that enables low maintenance also protects the motor and drive mechanism for longer service life. Building on more than 40 years of experience in shaker technology, Solaris™ Orbital Shakers are designed in Germany and manufactured in the United States, to deliver worry-free performance you can rely on. A full 10-year warranty is offered for the drive mechanism, plus a 5-year warranty on parts, and a two-year warranty on labor.

This is a compact, versatile, quiet benchtop companion. Solaris™ Orbital Shakers feature a small footprint that help save valuable benchtop space. Engineered to perform with a reduced noise level in a wide range of environments, units operate in a temperature range of 5 to +40 °C and humidity conditions from 20 to 80% non-condensing.

Certifications: UL, CE, and cUL (orbital shakers).

Ordering information: Solaris™ 2000 Series shakers ship with a 30.5×35.6 cm (12×14") universal platform. Solaris™ 4000 Series shakers ship with a 76.2×45.7 cm (18×30") universal platform. Additional platform sizes and dual stacks are also available.

Caution: 100/240 V models.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty