Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HyperSep™ Retain PEP SPE Plates

Supplier: Thermo Scientific
HyperSep™
60306-207 60303-207 60306-208
10048-506EA 575.99 USD InStock
10048-506 10048-474
HyperSep™ Retain PEP SPE Plates
Microplates Solid Phase Extraction Plates (SPE)
Ideal for large samples, Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation. Versatile polymeric material for retention of polar and non-polar analytes.

  • High and consistent recoveries
  • Fast and easy sample preparation and method development
  • pH stable zero to 14

HyperSep™ Retain™ PEP offers reproducible and reliable sample preparation through excellent  batch-to-batch reproducibility. Many of the problems associated with traditional silica-based materials, such as inconsistent recoveries due to sorbent drying, are eliminated when using HyperSep™ Retain™ PEP. This can lead to fast sample preparation and method development.

Particle size: 40 to 60 µm
Pore size: 55 to 90 Å
Surface area: 550 to 750 m²/g

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty