Would you like to log in again?
TraceGOLD™ TG-VVOC B GC Column
Supplier: Thermo Scientific
TraceGOLD™
26058-5180
76311-730EA 1999.26 USD InStock
76311-730
TraceGOLD™ TG-VVOC B GC Column
Chromatography Columns
Use Thermo Scientific™ TraceGOLD™ TG-VVOC B GC columns for unique selectivity of very volatile organic compounds (VVOC) like ozone precursors, air toxics, and volatile amines in environmental and industrial samples.
  • Highly inert, good retention, and efficiency
  • Great stability and chromatographic performance in difficult matrices
  • Highly robust with maximum temperature up to 290 °C
This highly inert gas chromatography (GC) capillary column, combined with its unique stable bonded phases, ensures excellent chromatographic peak shapes and great stability in difficult matrices such as water or ammonia.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty