Would you like to log in again?
Versaflo™ Single Station Battery Charger Kit, 3M™
Supplier: 3M
Versaflo™
7100035838
76312-268CS 396.71 USD InStock
76312-268
Versaflo™ Single Station Battery Charger Kit, 3M™
Batteries
This battery charger kit is a complete single station battery charger for TR-600 series PAPRs. It consists of a cradle and a power supply.
  • Easy to use
  • Quickly charging batteries
  • Holds charged batteries ready for use
  • Charging time: less than four hours
This battery charger will charge a standard capacity battery or high capacity battery in less than four hours. It will charge one battery at a time. Batteries may be stored on the charger, fully charged, ready for immediate use.
Certifications: NIOSH approved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty