Would you like to log in again?
Whatman™ Benchkote Sheets for ÄKTA Systems, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Benchkote® Whatman™ Benchkote®
2300-10065 2300-10062 2300-10064 2300-10092 2300-10074 2300-10093 2300-10074 2300-10063 2300-10060 2300-10060 2300-10065 2300-10061 2300-10073 2300-10094 2300-10064 2300-10092 2300-10072 2300-10061 2300-10063 2300-10073 2300-10072 2300-10093 2300-10094 2300-10062
76312-382PK 33.53 USD InStock
CA76312-390 CA76312-380 76312-382 CA76402-822 76312-372 76312-384 CA76312-382 CA76402-820 76402-820 76312-390 76312-380 76402-822 CA76312-376 CA76312-388 CA76312-372 76312-374 76402-818 CA76402-818 CA76312-384 76312-386 76312-376 CA76312-374 76312-388 CA76312-386
Whatman™ Benchkote Sheets for ÄKTA Systems, Whatman products (Cytiva)
Surface Protectors Bench Protectors
These Benchkote® sheets for ÄKTA systems will protect the top buffer trays from buffer spillages and salt deposits.
  • Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems
Benchkote® material has a high quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty