Would you like to log in again?
934 AH RTU MICROFIBRE FILTER 24 MM PK100
Supplier: Cytiva
934 Ah Rtu Microfibre Filter 24 Mm PK100^ Microfiber filter, Grade 934-AH* RTU, Material: Borosilicate glass membrane, One of the most widely used filters for Total Suspended Solids Analysis and is now available in a time saving Ready-To-Use (RTU) format, Binder-free, high purity
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
934 AH RTU MICROFIBRE FILTER 24 MM PK100 76312-392 9907-024 Pack of 100 (1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty