Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SMART Digest™ Protein Digests Kits

Supplier: Thermo Scientific
SMART Digest™
60109-101-LPH 60109-104-MB 60109-107 60109-101-MB 60109-108 60109-103-B 60109-101-B 60109-102-MB 60109-106 60109-102-B 60109-109-B 60109-107-MB 60109-105-MB 60109-101 60109-104-B 60109-105-B 60109-107-B 60109-102 60113-101 60109-103-MB 60109-106-B 60109101B2BUF 60109-109-MB 60109-108-MB 60109-106-MB 60109-104 60109-109 60109-108-B 60109-105 60109-103
75838-880EA 1132.03 USD InStock
75838-880 75839-428 75838-882 75839-404 75839-426 75838-884 75839-402 75839-564 75839-400 75838-888 75838-900 75838-902 75838-890 75838-870 75838-892 75838-894 75838-872 75838-874 75838-896 75838-876 75838-898 10017-750 75839-430 75839-574 75838-878 76312-780 10790-388 10790-386 10790-384 76005-236
SMART Digest™ Protein Digests Kits
Protein Extraction Protein Extraction Reagents
Obtain high quality analytical results from protein digests by using Thermo Scientific™ SMART Digest™ Kits.

  • Increased speed
  • Three-step sample preparation
  • High reproducibility
  • Higher sequence coverage
  • Compatible with automation

A significant advance in sample preparation for biopharmaceutical and protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation.

Significantly increased speed of sample processing with 3-step sample preparation. Significantly reduces sample preparation time, typically takes less than 60 minutes (dependant on sample complexity) to achieve full digestion. Higher sequence coverage is due to the removal of the reduction and alkylation steps and absence of trypsin. This reduces the number of chemicals and presence of trypsin in the final sample, as well as fewer post translational modifications, resulting in lower suppression of signal during LC-MS analysis.

For use with: Peptide quantitation, peptide mapping.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty