Would you like to log in again?
Tissue Plasminogen Activator (tPA) Activity Assay Kit (Colorimetric), BioVision
Supplier: BioVision
AB287878-100TEST
76312-782EA 867.85 USD InStock
76312-782
Tissue Plasminogen Activator (tPA) Activity Assay Kit (Colorimetric), BioVision
Assays Protein Assays
The most sensitive assay to measure tPA activity in biological samples.
  • Non-radioactive
  • Highthroughput compatible
  • Rapid, easy, one-step assay
  • Rapid assessment of enzymatic activity of tPA in biological samples
Tissue plasminogen activator (tPA, PLAT or tissue-type plasminogen activator; EC 3.4.21.68) is a serine protease that is found in blood and some tissues. tPA is the major intravascular activator of plasminogen. Thus, tPA is a key enzyme and the primary initiator of fibrinolysis in the vascular system where it plays an important role in the breakdown of blood clots. tPA has therapeutic uses and it is known as the 'clot buster' in clinical settings. High tPA activity in blood leads to hyperfibrinolysis and causes excessive bleeding; while low tPA activity leads to hypofibrinolysis, a condition that increases patient’s risks of suffering an arterial thrombosis.
BioVision’s tissue Plasminogen activator (tPA) activity assay kit of tPA utilizes the ability of active tPA to hydrolyze the synthetic substrate releasing pNA (chromophore). The released pNA can be easily quantified by OD: 405 nm. Our kit uses a unique combination of substrate and inhibitor mix that specifically detects tPA in a variety of biological samples. Other enzymes with similar catalytic properties [i.e. uPA (urokinase or urokinase-type plasminogen activator)] do not interfere with the assay. This assay kit is simple, specific and can detect as low as 5 µU of tPA activity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty